Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082491.4 | XP_001082491.4 | 790 | 63 | ||
XM_015119493.2 | XP_014974979.2 | 954 | 63 | |||
XM_015119494.2 | XP_014974980.1 | 2327 | 63 | |||
XM_015119500.2 | XP_014974986.1 | 488 | 63 | |||
XM_028836428.1 | XP_028692261.1 | 705 | 63 | |||
XM_028836429.1 | XP_028692262.1 | 913 | 63 | |||
XM_028836430.1 | XP_028692263.1 | 953 | 63 | |||
XM_028836431.1 | XP_028692264.1 | 851 | 63 | |||
GenBank mRNA | JU348927.1 | - | 442 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |