Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265644.1 | NP_001252573.1 | 413 | 153 | ||
GenBank mRNA | JV636440.1 | - | 413 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |