Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084619.4 | XP_001084619.2 | 1313 | 115 | ||
XM_028829264.1 | XP_028685097.1 | 1313 | 115 | |||
XM_028829265.1 | XP_028685098.1 | 1359 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |