Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084619.4 | XP_001084619.2 | 1457 | 76 | ||
GenBank mRNA | JU336522.1 | - | 1113 | 76 | ||
JU336523.1 | - | 1113 | 76 | |||
JU477331.1 | - | 1124 | 76 | |||
JU477332.1 | - | 1124 | 76 | |||
JV048325.1 | - | 1108 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |