Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091010.4 | XP_001091010.3 | 482 | 78 | ||
GenBank mRNA | JU479030.1 | - | 420 | 78 | ||
JV640579.1 | - | 383 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |