Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266098.1 | NP_001253027.1 | 1155 | 146 | ||
XM_015151468.2 | XP_015006954.1 | 1713 | 146 | |||
XM_028828851.1 | XP_028684684.1 | 1377 | 146 | |||
GenBank mRNA | JU321578.1 | - | 1530 | 146 | ||
JU321579.1 | - | 1526 | 146 | |||
JU472211.1 | - | 1706 | 146 | |||
JU472212.1 | - | 1702 | 146 | |||
JV045338.1 | - | 1155 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |