Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261302.1 | NP_001248231.1 | 688 | 97 | ||
XM_015115132.2 | XP_014970618.1 | 757 | 97 | |||
XM_028832855.1 | XP_028688688.1 | 848 | 97 | |||
GenBank mRNA | JU477357.1 | - | 688 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |