Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266728.2 | NP_001253657.1 | 1060 | 98 | ||
GenBank mRNA | JU322213.1 | - | 1047 | 98 | ||
JU471161.1 | - | 1060 | 98 | |||
JV638636.1 | - | 1069 | 98 |