Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194300.2 | NP_001181229.1 | 502 | 115 | ||
XM_015138967.2 | XP_014994453.1 | 554 | 115 | |||
GenBank mRNA | JV072776.1 | - | 240 | 115 | ||
JV635675.1 | - | 505 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |