Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086142.4 | XP_001086142.1 | 592 | 133 | ||
XM_028835853.1 | XP_028691686.1 | 586 | 133 | |||
XM_028835854.1 | XP_028691687.1 | 592 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |