Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194319.3 | NP_001181248.1 | 188 | 130 | ||
GenBank mRNA | JU332166.1 | - | 173 | 130 | ||
JU473602.1 | - | 120 | 130 | |||
JV044854.1 | - | 208 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |