Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842757.1 | XP_028698590.1 | 852 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |