Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091617.4 | XP_001091617.3 | 613 | 119 | ||
XM_015121973.2 | XP_014977459.1 | 618 | 119 | |||
XM_015121974.2 | XP_014977460.1 | 618 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |