Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261749.1 | NP_001248678.1 | 480 | 93 | ||
XM_015127698.2 | XP_014983184.1 | 578 | 93 | |||
XM_028841768.1 | XP_028697601.1 | 582 | 93 | |||
GenBank mRNA | JU474360.1 | - | 480 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |