Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266720.2 | NP_001253649.1 | 685 | 145 | ||
XM_015147634.2 | XP_015003120.1 | 737 | 145 | |||
XM_015147636.2 | XP_015003122.1 | 1036 | 145 | |||
XM_028853355.1 | XP_028709188.1 | 646 | 145 | |||
GenBank mRNA | JU322797.1 | - | 789 | 145 | ||
JU472827.1 | - | 778 | 145 | |||
JV045627.1 | - | 786 | 145 | |||
JV565064.1 | - | 685 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |