Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260951.1 | NP_001247880.1 | 83 | 117 | ||
GenBank mRNA | JU332899.1 | - | 70 | 117 | ||
JU474289.1 | - | 70 | 117 | |||
JV046171.1 | - | 83 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |