Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257951.1 | NP_001244880.1 | 283 | 137 | ||
XM_015118678.2 | XP_014974164.1 | 351 | 137 | |||
GenBank mRNA | JU322563.1 | - | 283 | 137 | ||
JU471401.1 | - | 193 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |