Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105068.4 | XP_001105068.1 | 219 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |