Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122280.2 | XP_014977766.2 | 343 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Macaca mulatta |