Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851309.1 | XP_028707142.1 | 5657 | 142 | ||
XM_028851310.1 | XP_028707143.1 | 5475 | 142 |
Gene Symbol: KIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU495902.1 | - | 3558 | 142 | ||
JU495903.1 | - | 3549 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |