Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT61A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257540.1 | NP_001244469.1 | 361 | 76 | ||
XM_015144519.2 | XP_015000005.1 | 507 | 76 | |||
GenBank mRNA | JU321217.1 | - | 361 | 76 | ||
JV044583.1 | - | 459 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |