Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090991.4 | XP_001090991.2 | 1561 | 126 | ||
XM_015151200.2 | XP_015006686.2 | 695 | 126 | |||
GenBank mRNA | JU479973.1 | - | 335 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |