Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC716404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836627.1 | XP_028692460.1 | 979 | 131 | ||
XM_028836628.1 | XP_028692461.1 | 496 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |