Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112371.2 | XP_014967857.1 | 2278 | 73 | ||
XM_015112372.2 | XP_014967858.1 | 1683 | 73 | |||
XM_028831672.1 | XP_028687505.1 | 2729 | 73 | |||
XM_028831673.1 | XP_028687506.1 | 1508 | 73 | |||
XM_028831674.1 | XP_028687507.1 | 1193 | 73 | |||
XM_028831675.1 | XP_028687508.1 | 963 | 73 | |||
GenBank mRNA | JU480348.1 | - | 564 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |