Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266134.2 | NP_001253063.1 | 719 | 125 | ||
XM_015132160.2 | XP_014987646.1 | 785 | 125 | |||
XM_015132162.2 | XP_014987648.1 | 773 | 125 | |||
XM_015132163.2 | XP_014987649.2 | 786 | 125 | |||
XM_015132165.1 | XP_014987651.1 | 655 | 125 | |||
XM_015132166.2 | XP_014987652.1 | 441 | 125 | |||
XM_015132167.2 | XP_014987653.1 | 786 | 125 | |||
XM_028843086.1 | XP_028698919.1 | 786 | 125 | |||
GenBank mRNA | JU323392.1 | - | 719 | 125 | ||
JU472818.1 | - | 759 | 125 | |||
JV044119.1 | - | 721 | 125 | |||
JV642439.1 | - | 774 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |