Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851517.1 | XP_028707350.1 | 2456 | 103 | ||
XM_028851518.1 | XP_028707351.1 | 1889 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |