Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028834552.1 | XP_028690385.1 | 470 | 55 | ||
XM_028834555.1 | XP_028690388.1 | 470 | 55 | |||
XM_028834557.1 | XP_028690390.1 | 470 | 55 | |||
XM_028834561.1 | XP_028690394.1 | 470 | 55 | |||
XM_028834567.1 | XP_028690400.1 | 470 | 55 | |||
XM_028834572.1 | XP_028690405.1 | 470 | 55 | |||
XM_028834574.1 | XP_028690407.1 | 470 | 55 | |||
XM_028834579.1 | XP_028690412.1 | 470 | 55 | |||
GenBank mRNA | JU474422.1 | - | 198 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |