Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142219.2 | XP_014997705.2 | 834 | 99 | ||
XM_028850897.1 | XP_028706730.1 | 765 | 99 | |||
XM_028850898.1 | XP_028706731.1 | 892 | 99 | |||
XM_028850899.1 | XP_028706732.1 | 823 | 99 | |||
XM_028850900.1 | XP_028706733.1 | 833 | 99 | |||
XM_028850901.1 | XP_028706734.1 | 833 | 99 |