Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266038.1 | NP_001252967.1 | 433 | 122 | ||
XM_028837739.1 | XP_028693572.1 | 480 | 122 | |||
GenBank mRNA | JU320219.1 | - | 433 | 122 | ||
JV574491.1 | - | 472 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |