Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H9orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101033.4 | XP_001101033.1 | 1212 | 144 | ||
XM_015117039.2 | XP_014972525.1 | 1059 | 144 | |||
XM_028834963.1 | XP_028690796.1 | 1793 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |