Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838295.1 | XP_028694128.1 | 1115 | 93 | ||
XM_028838296.1 | XP_028694129.1 | 1122 | 93 | |||
XM_028838297.1 | XP_028694130.1 | 918 | 93 | |||
GenBank mRNA | JU333070.1 | - | 887 | 93 | ||
JV045386.1 | - | 862 | 93 | |||
JV673909.1 | - | 898 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |