Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193989.2 | NP_001180918.1 | 1154 | 132 | ||
XM_015120901.2 | XP_014976387.1 | 1238 | 132 | |||
XM_028837113.1 | XP_028692946.1 | 1230 | 132 | |||
XM_028837114.1 | XP_028692947.1 | 704 | 132 | |||
GenBank mRNA | JU323204.1 | - | 1139 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |