Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194032.2 | NP_001180961.1 | 378 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |