Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194174.1 | NP_001181103.1 | 156 | 108 | ||
GenBank mRNA | JU321940.1 | - | 188 | 108 | ||
JU473334.1 | - | 156 | 108 | |||
JV046447.1 | - | 168 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |