Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257556.1 | NP_001244485.1 | 497 | 118 | ||
GenBank mRNA | JU320923.1 | - | 497 | 118 | ||
JU473059.1 | - | 473 | 118 | |||
JV646005.1 | - | 95 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |