Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086413.4 | XP_001086413.1 | 3173 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |