Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828765.1 | XP_028684598.1 | 1249 | 148 | ||
XM_028828766.1 | XP_028684599.1 | 1563 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |