Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258194.1 | NP_001245123.1 | 1420 | 130 | ||
GenBank mRNA | JU333316.1 | - | 1363 | 130 | ||
JU474625.1 | - | 1420 | 130 | |||
JV046404.1 | - | 1338 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |