Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090704.3 | XP_001090704.1 | 702 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |