Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171840.1 | NP_001165311.1 | 310 | 135 | ||
XM_028834337.1 | XP_028690170.1 | 577 | 135 | |||
XM_028834338.1 | XP_028690171.1 | 369 | 135 | |||
GenBank mRNA | AF218048.1 | - | 310 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |