Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123900.2 | XP_014979386.1 | 3063 | 85 | ||
XM_015123909.2 | XP_014979395.1 | 3204 | 85 | |||
XM_015123916.2 | XP_014979402.1 | 3110 | 85 | |||
XM_015123920.2 | XP_014979406.1 | 2970 | 85 | |||
XM_028850547.1 | XP_028706380.1 | 3195 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |