Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105766.4 | XP_001105766.1 | 869 | 92 | ||
XM_028833954.1 | XP_028689787.1 | 736 | 92 |