Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143175.2 | XP_014998661.1 | 981 | 106 | ||
XM_015143176.2 | XP_014998662.1 | 1226 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |