Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266344.1 | NP_001253273.1 | 1285 | 149 | ||
GenBank mRNA | JU333211.1 | - | 1285 | 149 | ||
JV703816.1 | - | 1529 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |