Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261373.1 | NP_001248302.1 | 514 | 136 | ||
XM_015152564.2 | XP_015008050.1 | 576 | 136 | |||
XM_015152577.2 | XP_015008063.1 | 582 | 136 | |||
XM_015152587.2 | XP_015008073.1 | 582 | 136 | |||
XM_015152597.2 | XP_015008083.1 | 838 | 136 | |||
XM_028845384.1 | XP_028701217.1 | 838 | 136 | |||
XM_028845386.1 | XP_028701219.1 | 402 | 136 | |||
XM_028845391.1 | XP_028701224.1 | 838 | 136 | |||
GenBank mRNA | JU333300.1 | - | 470 | 136 | ||
JU474613.1 | - | 511 | 136 | |||
JV046393.1 | - | 514 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |