Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125015.2 | XP_014980501.1 | 537 | 90 | ||
XM_015125016.2 | XP_014980502.1 | 906 | 90 | |||
XM_028840063.1 | XP_028695896.1 | 528 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |