Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194458.2 | NP_001181387.1 | 592 | 80 | ||
GenBank mRNA | JU333023.1 | - | 592 | 80 | ||
JU333024.1 | - | 592 | 80 | |||
JV571823.1 | - | 597 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |