Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097091.4 | XP_001097091.1 | 332 | 97 | ||
XM_001097205.4 | XP_001097205.1 | 288 | 97 | |||
XM_028834614.1 | XP_028690447.1 | 333 | 97 | |||
XM_028834615.1 | XP_028690448.1 | 522 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |