Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096810.4 | XP_001096810.2 | 887 | 93 | ||
XM_015147579.2 | XP_015003065.2 | 1292 | 93 | |||
XM_028826616.1 | XP_028682449.1 | 1166 | 93 | |||
XM_028826617.1 | XP_028682450.1 | 1217 | 93 | |||
XM_028826618.1 | XP_028682451.1 | 1139 | 93 | |||
XM_028826619.1 | XP_028682452.1 | 1113 | 93 | |||
GenBank mRNA | JU324011.1 | - | 589 | 93 | ||
JV645225.1 | - | 642 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |