Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089204.4 | XP_001089204.1 | 355 | 146 | ||
XM_015144766.2 | XP_015000252.1 | 447 | 146 | |||
XM_028852333.1 | XP_028708166.1 | 448 | 146 | |||
XM_028852336.1 | XP_028708169.1 | 444 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |